missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-nitrobenzoate, 98%

Catalog No. AAA1532906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1532906 5 g
AAA1532914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1532906 Supplier Thermo Scientific Chemicals Supplier No. A1532906

CAS: 713-52-0 | C8H7NO5 | 197.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 713-52-0
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00016997
InChI Key UEGCRFNWTGYVKX-UHFFFAOYSA-N
Synonym benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
PubChem CID 294866
IUPAC Name methyl 3-hydroxy-4-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 89°C to 92°C
Quantity 5 g
Beilstein 2583832
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only