missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxy-2-thiophenecarboxylate 97.0+%, TCI America™

Catalog No. M27165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27165G 5 g
M27161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27165G Supplier TCI America Supplier No. M27165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5118-06-9
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.17
MDL Number MFCD00055642
InChI Key FYOPNADYSPZUAT-WAYWQWQTSA-N
Synonym methyl 3-hydroxy-2-thiophenecarboxylate, 3-hydroxy-2-thiophenecarboxylic acid methyl ester, 3-hydroxy-thiophene-2-carboxylic acid methyl ester, 3-hydroxythiophene-2-carboxylic acid methyl ester, pubchem7744, acmc-1aqhj, maybridge3_005989, 3-hydroxy-2-methoxycarbonylthiophene, 3-hydroxy-2-methoxycarbonyl-thiophene, methyl 3-hydroxythiophen-2-caboxylate
PubChem CID 581127
IUPAC Name methyl 3-hydroxythiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)O

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 158.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxy-2-thiophenecarboxylate