accessibility menu, dialog, popup

UserName

CAS RN 5118-06-9

IUPAC Name: (2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Synonyms: (2Z)-2-[hydroxy(methoxy)methylidene]thiophen-3-one
Molecular Weight (g/mol): 158.17
Molecular Formula: C6H6O3S
InChi Key: FYOPNADYSPZUAT-WAYWQWQTSA-N
SMILES: CO\C(O)=C1/SC=CC1=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)