missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-2-naphthoate, 98+%

Catalog No. AAL1453306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1453306 5 g
AAL1453314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1453306 Supplier Thermo Scientific Chemicals Supplier No. L1453306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 883-99-8 | C12H10O3 | 202.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 883-99-8
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004099
InChI Key YVVBECLPRBAATK-UHFFFAOYSA-N
Synonym methyl 3-hydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 3-hydroxy-, methyl ester, methyl 2-hydroxy-3-naphthoate, 2-hydroxy-3-naphthoic acid methyl ester, 3-hydroxy-2-naphthoic acid methyl ester, methyl 3-hydroxy-2-naphthalenecarboxylate, 2-naphthoic acid, 3-hydroxy-, methyl ester, methyl3-hydroxy-2-naphthoate, acmc-209qsx, 3-methoxycarbonyl-2-naphthol
PubChem CID 70163
IUPAC Name methyl 3-hydroxynaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2C=C1O

Specifications

Melting Point 72°C to 74°C
Boiling Point 205°C to 207°C (160 mmHg)
Quantity 5 g
Beilstein 1074526
Formula Weight 202.21
Percent Purity ≥98%
Chemical Name or Material Methyl 3-hydroxy-2-naphthoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-936-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only