missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Furancarboxylate 96.0+%, TCI America™

Catalog No. F04685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04685G 5 g
F046825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04685G Supplier TCI America Supplier No. F04685G
Only null left

Chemical Identifiers

CAS 13129-23-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD06203671
InChI Key ZKHQSQYLKSSYIP-UHFFFAOYSA-N
Synonym 3-Furoic Acid Methyl Ester, 3-Furancarboxylic Acid Methyl Ester, Methyl 3-Furoate
PubChem CID 14918
IUPAC Name methyl furan-3-carboxylate
SMILES COC(=O)C1=COC=C1

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
UN Number 3272
Formula Weight 126.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Furancarboxylate