accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 13129-23-2

IUPAC Name: methyl furan-3-carboxylate
Synonyms: methyl furan-3-carboxylate
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: ZKHQSQYLKSSYIP-UHFFFAOYSA-N
SMILES: COC(=O)C1=COC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results