missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Fluorophenylacetate 98.0+%, TCI America™

Catalog No. M25365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25365G 5 g
M253625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25365G Supplier TCI America Supplier No. M25365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64123-77-9
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD03093288
InChI Key OXYLCGRXCQARQV-UHFFFAOYSA-N
PubChem CID 2734108
IUPAC Name methyl 2-(3-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(F)=C1

Specifications

Color Colorless
Quantity 5 g
Synonym methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
Formula Weight 168.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Fluorophenylacetate