missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluoro-4-methoxybenzoate, 95%, Thermo Scientific™

Catalog No. AAH3199409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3199409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3199409 Supplier Thermo Scientific Chemicals Supplier No. H3199409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 369-30-2
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00215839
InChI Key MIKOKYAREMINFF-UHFFFAOYSA-N
Synonym benzoic acid, 3-fluoro-4-methoxy-, methyl ester, methyl 3-fluoro-4-methyloxy benzoate, 3-fluoro-4-methoxybenzoic acid methyl ester, pubchem3569, acmc-1ck9v, rarechem al bf 0055, #
PubChem CID 592848
IUPAC Name methyl 3-fluoro-4-methoxybenzoate
SMILES COC(=O)C1=CC(F)=C(OC)C=C1

Specifications

Melting Point 69°C to 73°C
Quantity 10 g
Formula Weight 184.17
Percent Purity 95%
Chemical Name or Material Methyl 3-fluoro-4-methoxybenzoate
TSCA No
Recommended Storage Ambient temperatures