missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Fluoro-2-thiophenecarboxylate 97.0+%, TCI America™

Catalog No. m22951g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22951G Supplier TCI America Supplier No. M22951G
Only null left

Chemical Identifiers

CAS 100421-52-1
Molecular Formula C6H5FO2S
Molecular Weight (g/mol) 160.162
MDL Number MFCD16606491
InChI Key UKIUEFZYSXIGPY-UHFFFAOYSA-N
PubChem CID 10583081
IUPAC Name methyl 3-fluorothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)F

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 1 g
Formula Weight 160.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Fluoro-2-thiophenecarboxylate
TSCA No
Recommended Storage Refrigerator