accessibility menu, dialog, popup

UserName

CH3OOSF

CAS RN 100421-52-1

IUPAC Name: methyl 3-fluorothiophene-2-carboxylate
Synonyms: methyl 3-fluorothiophene-2-carboxylate
Molecular Weight (g/mol): 160.16
Molecular Formula: C6H5FO2S
InChi Key: UKIUEFZYSXIGPY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results