missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-ethoxythiophene-2-carboxylate, 97%

CAS: 139926-22-0 | C8H10O3S | 186.23 g/mol

Supplier:  Thermo Scientific Chemicals B2086006

 View more versions of this product

Catalog No. AAB2086006

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 3-ethoxythiophene-2-carboxylate
72°C to 74°C
MFCD00102116
maybridge1_003754, acmc-1c0r9, methyl-3-ethoxythiophene carboxylate, 3-ethoxy-2-thiophenecarboxylic acid methyl ester, 3-ethoxythiophene-2-carboxylic acid methyl ester, 2,3-ethoxy-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-ethoxy-, methyl ester
CCOC1=C(SC=C1)C(=O)OC
186.23
186.23
139926-22-0
C8H10O3S
5 g
JAGLXKOXARHKBR-UHFFFAOYSA-N
methyl 3-ethoxythiophene-2-carboxylate
2777650
97%

Chemical Identifiers

139926-22-0
186.23
JAGLXKOXARHKBR-UHFFFAOYSA-N
2777650
CCOC1=C(SC=C1)C(=O)OC
C8H10O3S
MFCD00102116
maybridge1_003754, acmc-1c0r9, methyl-3-ethoxythiophene carboxylate, 3-ethoxy-2-thiophenecarboxylic acid methyl ester, 3-ethoxythiophene-2-carboxylic acid methyl ester, 2,3-ethoxy-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-ethoxy-, methyl ester
methyl 3-ethoxythiophene-2-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only