missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-ethoxythiophene-2-carboxylate, 97%

Catalog No. AAB2086003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2086003 1 g
AAB2086006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2086003 Supplier Thermo Scientific Chemicals Supplier No. B2086003

CAS: 139926-22-0 | C8H10O3S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139926-22-0
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00102116
InChI Key JAGLXKOXARHKBR-UHFFFAOYSA-N
Synonym maybridge1_003754, acmc-1c0r9, methyl-3-ethoxythiophene carboxylate, 3-ethoxy-2-thiophenecarboxylic acid methyl ester, 3-ethoxythiophene-2-carboxylic acid methyl ester, 2,3-ethoxy-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-ethoxy-, methyl ester
PubChem CID 2777650
IUPAC Name methyl 3-ethoxythiophene-2-carboxylate
SMILES CCOC1=C(SC=C1)C(=O)OC

Specifications

Melting Point 72°C to 74°C
Quantity 1 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Methyl 3-ethoxythiophene-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only