missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%

Catalog No. p-4510691
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC372020010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC372020010 Supplier Thermo Scientific Chemicals Supplier No. 372020010
Only null left

CAS: 113212-14-9 | C7H10N2O2 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113212-14-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
InChI Key FREGRQYTSPAJDX-AATRIKPKSA-N
Synonym 3-dimethylamino-2-isocyanoacrylic acid methyl ester, methyl e-3-dimethylamino-2-isocyanoprop-2-enoate, methyl 2e-3-dimethylamino-2-isocyanoprop-2-enoate
PubChem CID 6387542
IUPAC Name methyl (E)-3-(dimethylamino)-2-isocyanoprop-2-enoate
SMILES CN(C)C=C(C(=O)OC)[N+]#[C-]

Specifications

Melting Point 61.0°C to 67.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 154.17
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-(dimethylamino)-2-isocyanoacrylate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only