missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chloro-5-iodobenzoate, 98%, Thermo Scientific Chemicals

$785.56 - $785.56

Chemical Identifiers

CAS 289039-85-6
Molecular Formula C8H6ClIO2
Molecular Weight (g/mol) 296.488
MDL Number MFCD00673018
InChI Key ZDSIXLJPASGHDW-UHFFFAOYSA-N
Synonym acmc-20akhf, methyl 5-chloro-3-iodobenzoate, 3-chloro-5-iodo-benzoic acid methyl ester, 3-chloro-5-iodobenzoic acid methyl ester
PubChem CID 24721134
IUPAC Name methyl 3-chloro-5-iodobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)I)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2544406
View Documents
Thermo Scientific Chemicals
B2544406
5 g
Each for $785.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

289039-85-6
296.488
ZDSIXLJPASGHDW-UHFFFAOYSA-N
24721134
COC(=O)C1=CC(=CC(=C1)I)Cl
C8H6ClIO2
MFCD00673018
acmc-20akhf, methyl 5-chloro-3-iodobenzoate, 3-chloro-5-iodo-benzoic acid methyl ester, 3-chloro-5-iodobenzoic acid methyl ester
methyl 3-chloro-5-iodobenzoate

Specifications

289039-85-6
C8H6ClIO2
5 g
acmc-20akhf, methyl 5-chloro-3-iodobenzoate, 3-chloro-5-iodo-benzoic acid methyl ester, 3-chloro-5-iodobenzoic acid methyl ester
COC(=O)C1=CC(=CC(=C1)I)Cl
296.488
296.49
Methyl 3-chloro-5-iodobenzoate
41°C to 43°C
MFCD00673018
Light sensitive
ZDSIXLJPASGHDW-UHFFFAOYSA-N
methyl 3-chloro-5-iodobenzoate
24721134
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only