missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(Bromomethyl)benzoate 97.0+%, TCI America™
$90.48 - $273.38
Chemical Identifiers
| CAS | 1129-28-8 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.07 |
| MDL Number | MFCD00051437 |
| InChI Key | YUHSMQQNPRLEEJ-UHFFFAOYSA-N |
| PubChem CID | 517981 |
| IUPAC Name | methyl 3-(bromomethyl)benzoate |
| SMILES | COC(=O)C1=CC=CC(CBr)=C1 |
Chemical Identifiers
| 1129-28-8 | |
| 229.07 | |
| YUHSMQQNPRLEEJ-UHFFFAOYSA-N | |
| methyl 3-(bromomethyl)benzoate |
| C9H9BrO2 | |
| MFCD00051437 | |
| 517981 | |
| COC(=O)C1=CC=CC(CBr)=C1 |
Specifications
| 1129-28-8 | |
| 169°C | |
| MFCD00051437 | |
| 3261 | |
| YUHSMQQNPRLEEJ-UHFFFAOYSA-N | |
| methyl 3-(bromomethyl)benzoate | |
| 517981 | |
| ≥97.0% (GC) | |
| Methyl 3-(Bromomethyl)benzoate |
| 44°C | |
| C9H9BrO2 | |
| 5 g | |
| methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide | |
| COC(=O)C1=CC=CC(CBr)=C1 | |
| 229.07 | |
| 229.07 | |
| Crystalline Powder |
Safety and Handling
TSCA : No