missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(bromomethyl)benzoate, 97%

Catalog No. p-7081731
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAA1182606 5 g
AAA1182614 25 g
2 options

Catalog No. AAA1182606

Supplier: Thermo Scientific Chemicals A1182606

Only null left

CAS: 1129-28-8 | C9H9BrO2 | 229.07 g/mol

Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Keep away from strong bases and oxidizing agents. Light sensitive. Incompatible with reducing agents, amines.

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1

Specifications

Melting Point 43°C to 45°C
Boiling Point 115°C (0.8 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 638569
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl 3- (bromomethyl)benzoate
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only