missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(bromomethyl)benzoate, 97%

Catalog No. AAA1182614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1182614 25 g
AAA1182606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1182614 Supplier Thermo Scientific Chemicals Supplier No. A1182614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1129-28-8 | C9H9BrO2 | 229.07 g/mol

Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Keep away from strong bases and oxidizing agents. Light sensitive. Incompatible with reducing agents, amines.

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1

Specifications

Melting Point 43°C to 45°C
Boiling Point 115°C (0.8 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 638569
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl 3- (bromomethyl)benzoate
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only