missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(bromomethyl)benzoate, 95%

CAS: 1129-28-8 | C9H9BrO2 | 229.07 g/mol

$182.06 - $182.06

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368630050
SDS View Documents
Thermo Scientific Chemicals
368630050
5 g Glass bottle
Each for $182.06
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1129-28-8
229.07
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
517981
COC(=O)C1=CC=CC(CBr)=C1
C9H9BrO2
MFCD00051437
methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
methyl 3-(bromomethyl)benzoate

Specifications

1129-28-8
White
>110°C
94% min. (GC)
C9H9BrO2
MFCD00051437
09, IV, 1721
Solubility in water: .. Other solubilities: soluble in methanol
COC(=O)C1=CC=CC(CBr)=C1
229.07
229
Crystalline Powder or Crystals
35.0°C to 37.0°C
135.0°C (2.0 mmHg)
Authentic
Glass bottle
CH2BrC6H4C(O)OCH3
5 g
methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
methyl 3-(bromomethyl)benzoate
517981
95%
Methyl 3-(bromomethyl)benzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only