missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromobenzoate 99.0+%, TCI America™

Supplier:  TCI America B05545G

 View more versions of this product

Catalog No. B05545G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl 3-Bromobenzoate
White-Yellow
MFCD00017777
methyl m-bromobenzoate, 3-bromobenzoic acid methyl ester, benzoic acid, 3-bromo-, methyl ester, 3-bromobenzoic acid, methyl ester, methyl-3-bromobenzoate, benzoic acid, m-bromo-, methyl ester, m-bromobenzoic acid methyl ester, pubchem3961, methyl m-bromo benzoate, methyl 3-bromo-benzoate
COC(=O)C1=CC(=CC=C1)Br
215.046
215.05
Crystals
618-89-3
C8H7BrO2
5 g
KMFJVYMFCAIRAN-UHFFFAOYSA-N
methyl 3-bromobenzoate
12070
≥99.0% (GC)

Chemical Identifiers

618-89-3
215.046
KMFJVYMFCAIRAN-UHFFFAOYSA-N
12070
COC(=O)C1=CC(=CC=C1)Br
C8H7BrO2
MFCD00017777
methyl m-bromobenzoate, 3-bromobenzoic acid methyl ester, benzoic acid, 3-bromo-, methyl ester, 3-bromobenzoic acid, methyl ester, methyl-3-bromobenzoate, benzoic acid, m-bromo-, methyl ester, m-bromobenzoic acid methyl ester, pubchem3961, methyl m-bromo benzoate, methyl 3-bromo-benzoate
methyl 3-bromobenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions