missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromobenzoate, 98+%

Catalog No. AAA1617409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1617409 10 g
AAA1617418 50 g
AAA1617430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1617409 Supplier Thermo Scientific Chemicals Supplier No. A1617409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 618-89-3 | C8H7BrO2 | 215.046 g/mol

Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromobenzoate is used as a reagent in organic synthesis. It is used in the synthesis of tetrahydro dibenzazocine, which acts as an inhibitor of 17beta-hydroxysteroid dehydrogenase type 3. Further, it is used to prepare 3-tert-butylphosphanyl-benzoic acid methyl ester by reacting with tert-butyltrimethylsilylphosphine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 618-89-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00017777
InChI Key KMFJVYMFCAIRAN-UHFFFAOYSA-N
Synonym methyl m-bromobenzoate, 3-bromobenzoic acid methyl ester, benzoic acid, 3-bromo-, methyl ester, 3-bromobenzoic acid, methyl ester, methyl-3-bromobenzoate, benzoic acid, m-bromo-, methyl ester, m-bromobenzoic acid methyl ester, pubchem3961, methyl m-bromo benzoate, methyl 3-bromo-benzoate
PubChem CID 12070
IUPAC Name methyl 3-bromobenzoate
SMILES COC(=O)C1=CC(=CC=C1)Br

Specifications

Melting Point 30°C to 33°C
Boiling Point 139°C to 140°C (35 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5575
Quantity 10 g
Beilstein 1940810
Solubility Information Slightly soluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material Methyl 3-bromobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-569-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only