missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromobenzoate, 98%

Catalog No. p-161002
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC333650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC333650050 Supplier Thermo Scientific Chemicals Supplier No. 333650050

CAS: 618-89-3 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-89-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00017777
InChI Key KMFJVYMFCAIRAN-UHFFFAOYSA-N
Synonym methyl m-bromobenzoate, 3-bromobenzoic acid methyl ester, benzoic acid, 3-bromo-, methyl ester, 3-bromobenzoic acid, methyl ester, methyl-3-bromobenzoate, benzoic acid, m-bromo-, methyl ester, m-bromobenzoic acid methyl ester, pubchem3961, methyl m-bromo benzoate, methyl 3-bromo-benzoate
PubChem CID 12070
IUPAC Name methyl 3-bromobenzoate
SMILES COC(=O)C1=CC(=CC=C1)Br

Specifications

Color White to Yellow
Boiling Point 139°C to 140°C (35.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 215.05
Percent Purity 98%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material Methyl 3-bromobenzoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-569-9
TSCA TSCA

RUO – Research Use Only