missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Bromo-2-fluorobenzoate 98.0+%, TCI America™
$83.89 - $335.52
Chemical Identifiers
| CAS | 206551-41-9 |
|---|---|
| Molecular Formula | C8H6BrFO2 |
| Molecular Weight (g/mol) | 233.036 |
| MDL Number | MFCD09261256 |
| InChI Key | ZWOFHFOFKBYRHV-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester |
| PubChem CID | 21584505 |
| IUPAC Name | methyl 3-bromo-2-fluorobenzoate |
| SMILES | COC(=O)C1=C(C(=CC=C1)Br)F |
Chemical Identifiers
| 206551-41-9 | |
| 233.036 | |
| ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
| 21584505 | |
| COC(=O)C1=C(C(=CC=C1)Br)F |
| C8H6BrFO2 | |
| MFCD09261256 | |
| 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
| methyl 3-bromo-2-fluorobenzoate |
Specifications
| 206551-41-9 | |
| White | |
| MFCD09261256 | |
| 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
| COC(=O)C1=C(C(=CC=C1)Br)F | |
| 233.036 | |
| 233.04 | |
| Crystalline Powder |
| 37°C | |
| C8H6BrFO2 | |
| 1 g | |
| ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
| methyl 3-bromo-2-fluorobenzoate | |
| 21584505 | |
| ≥98.0% (GC) | |
| Methyl 3-Bromo-2-fluorobenzoate |
Safety and Handling
TSCA : No