missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-2-fluorobenzoate 98.0+%, TCI America™

Catalog No. M27115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27115G 5 g
M27111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27115G Supplier TCI America Supplier No. M27115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 206551-41-9
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD09261256
InChI Key ZWOFHFOFKBYRHV-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester
PubChem CID 21584505
IUPAC Name methyl 3-bromo-2-fluorobenzoate
SMILES COC(=O)C1=C(C(=CC=C1)Br)F

Specifications

Melting Point 37°C
Color White
Quantity 5 g
Formula Weight 233.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Bromo-2-fluorobenzoate