missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Bromo-2-fluorobenzoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 37°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 233.04 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 3-Bromo-2-fluorobenzoate |
Safety and Handling
| TSCA | No |