missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-bromo-2-fluorobenzoate, 98%, Thermo Scientific Chemicals
$457.22 - $457.22
Chemical Identifiers
| CAS | 206551-41-9 |
|---|---|
| Molecular Formula | C8H6BrFO2 |
| Molecular Weight (g/mol) | 233.036 |
| MDL Number | MFCD09261255 |
| InChI Key | ZWOFHFOFKBYRHV-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester |
| PubChem CID | 21584505 |
| IUPAC Name | methyl 3-bromo-2-fluorobenzoate |
| SMILES | COC(=O)C1=C(C(=CC=C1)Br)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3686506
|
Thermo Scientific Chemicals
H3686506 |
5 g |
Each for $457.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 206551-41-9 | |
| 233.036 | |
| ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
| 21584505 | |
| COC(=O)C1=C(C(=CC=C1)Br)F |
| C8H6BrFO2 | |
| MFCD09261255 | |
| 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
| methyl 3-bromo-2-fluorobenzoate |
Specifications
| 206551-41-9 | |
| White to Cream | |
| MFCD09261255 | |
| 3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
| COC(=O)C1=C(C(=CC=C1)Br)F | |
| 233.036 | |
| 233.04 | |
| Fused Solid |
| 35°C to 37°C | |
| C8H6BrFO2 | |
| 5 g | |
| ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
| methyl 3-bromo-2-fluorobenzoate | |
| 21584505 | |
| 98% | |
| Methyl 3-bromo-2-fluorobenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only