Learn More
Methyl 3-aminothiophene-4-carboxylate hydrochloride, 97+%
CAS: 39978-14-8 | C6H7NO2S | 157.19 g/mol
$162.58 - $658.47
Chemical Identifiers
| CAS | 39978-14-8 |
|---|---|
| Molecular Formula | C6H7NO2S |
| Molecular Weight (g/mol) | 157.19 |
| MDL Number | MFCD00068149 |
| InChI Key | BUFZZXCVOFBHLS-UHFFFAOYSA-N |
| Synonym | methyl 4-aminothiophene-3-carboxylate hydrochloride, methyl 3-aminothiophene-4-carboxylate hydrochloride, 3-carbomethoxy-4-aminothiophene hydrochloride, acmc-209zkz, methyl 4-aminothiophene-3-carboxylate, chloride, 3-amino-4-methoxycarbonylthiophene hydrochloride, 4-amino-3-methoxycarbonylthiophene hydrochloride, methyl3-aminothiophene-4-carboxylatehydrochloride, 4-amino-3-methoxycarbonyl thiophene hydrochloride, methyl 4-aminothiophen-3-carboxylate hydrochloride |
| PubChem CID | 2777609 |
| SMILES | COC(=O)C1=CSC=C1N |
Description
Methyl 3-aminothiophene-4-carboxylate hydrochloride is used in the preparation of ethyl (4-methoxycarbonyl)thiophene-3-carbamate by reacting with carbonochloridic acid ethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39978-14-8 | |
| 157.19 | |
| BUFZZXCVOFBHLS-UHFFFAOYSA-N | |
| 2777609 |
| C6H7NO2S | |
| MFCD00068149 | |
| methyl 4-aminothiophene-3-carboxylate hydrochloride, methyl 3-aminothiophene-4-carboxylate hydrochloride, 3-carbomethoxy-4-aminothiophene hydrochloride, acmc-209zkz, methyl 4-aminothiophene-3-carboxylate, chloride, 3-amino-4-methoxycarbonylthiophene hydrochloride, 4-amino-3-methoxycarbonylthiophene hydrochloride, methyl3-aminothiophene-4-carboxylatehydrochloride, 4-amino-3-methoxycarbonyl thiophene hydrochloride, methyl 4-aminothiophen-3-carboxylate hydrochloride | |
| COC(=O)C1=CSC=C1N |
Specifications
| 39978-14-8 | |
| Brown | |
| MFCD00068149 | |
| Hygroscopic | |
| BUFZZXCVOFBHLS-UHFFFAOYSA-N | |
| methyl 4-aminothiophene-3-carboxylate | |
| 2777609 | |
| ≥97% | |
| Methyl 3-aminothiophene-4-carboxylate hydrochloride |
| 199°C to 200°C (decomposition) | |
| C6H7NO2S | |
| 1 g | |
| methyl 4-aminothiophene-3-carboxylate hydrochloride, methyl 3-aminothiophene-4-carboxylate hydrochloride, 3-carbomethoxy-4-aminothiophene hydrochloride, acmc-209zkz, methyl 4-aminothiophene-3-carboxylate, chloride, 3-amino-4-methoxycarbonylthiophene hydrochloride, 4-amino-3-methoxycarbonylthiophene hydrochloride, methyl3-aminothiophene-4-carboxylatehydrochloride, 4-amino-3-methoxycarbonyl thiophene hydrochloride, methyl 4-aminothiophen-3-carboxylate hydrochloride | |
| COC(=O)C1=CSC=C1N | |
| 157.19 | |
| 193.65 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only