accessibility menu, dialog, popup

UserName

CAS RN 39978-14-8

IUPAC Name: methyl 4-aminothiophene-3-carboxylate
Synonyms: methyl 4-aminothiophene-3-carboxylate
Molecular Weight (g/mol): 157.19
Molecular Formula: C6H7NO2S
InChi Key: BUFZZXCVOFBHLS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CSC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results