missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-5-bromo-2-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7133468
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28441G 1 g
M28445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28441G Supplier TCI America Supplier No. M28441G
Only null left

Chemical Identifiers

CAS 1000342-11-9
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD08690071
InChI Key NMLOSXSDLWFBKT-UHFFFAOYSA-N
Synonym 3-Amino-5-bromo-2-methylbenzoic Acid Methyl Ester
PubChem CID 24729181
IUPAC Name methyl 3-amino-5-bromo-2-methylbenzoate
SMILES COC(=O)C1=CC(Br)=CC(N)=C1C

Specifications

Melting Point 52°C
Quantity 1 g
Formula Weight 244.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Amino-5-bromo-2-methylbenzoate