missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-4-methylbenzoate 98.0+%, TCI America™

Catalog No. A143225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A143225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A143225G Supplier TCI America Supplier No. A143225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18595-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00025206
InChI Key YEPWCJHMSVABPQ-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930
PubChem CID 337778
IUPAC Name methyl 3-amino-4-methylbenzoate
SMILES COC(=O)C1=CC=C(C)C(N)=C1

Specifications

Melting Point 116°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Amino-4-methylbenzoate