missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methylbenzoate, 97%

Catalog No. p-7022525
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1534806 5 g
AAA1534814 25 g
AAA1534822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1534806 Supplier Thermo Scientific Chemicals Supplier No. A1534806
Only null left

CAS: 18595-18-1 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18595-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00025206
InChI Key YEPWCJHMSVABPQ-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930
PubChem CID 337778
IUPAC Name methyl 3-amino-4-methylbenzoate
SMILES COC(=O)C1=CC=C(C)C(N)=C1

Specifications

Melting Point 113°C to 117°C
Quantity 5 g
Beilstein 2088611
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only