missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Amino-4-methoxybenzoate 98.0+%, TCI America™
Supplier: TCI America M17155G
Specifications
| Methyl 3-Amino-4-methoxybenzoate | |
| 86°C | |
| C9H11NO3 | |
| 3-amino-4-methoxybenzoic acid methyl ester, 3-amino-4-methoxy-benzoic acid methyl ester, methyl-3-amino-4-methoxybenzoate, 3-amino-p-anisic acid methyl ester, methyl-4-methoxy-3-aminobenzoate, benzoic acid, 3-amino-4-methoxy-, methyl ester, pubchem2033, 3-amino-4-methoxy-benzoicacidmethylester, acmc-209get, m-aminoanissauremethylester | |
| COC1=C(C=C(C=C1)C(=O)OC)N | |
| 181.191 | |
| 181.19 | |
| Crystalline Powder |
| 24812-90-6 | |
| 183°C | |
| 5 g | |
| QVDWKLDUBSJEOG-UHFFFAOYSA-N | |
| methyl 3-amino-4-methoxybenzoate | |
| 2734814 | |
| ≥98.0% (T) |
Chemical Identifiers
| 24812-90-6 | |
| 181.191 | |
| 3-amino-4-methoxybenzoic acid methyl ester, 3-amino-4-methoxy-benzoic acid methyl ester, methyl-3-amino-4-methoxybenzoate, 3-amino-p-anisic acid methyl ester, methyl-4-methoxy-3-aminobenzoate, benzoic acid, 3-amino-4-methoxy-, methyl ester, pubchem2033, 3-amino-4-methoxy-benzoicacidmethylester, acmc-209get, m-aminoanissauremethylester | |
| methyl 3-amino-4-methoxybenzoate |
| C9H11NO3 | |
| QVDWKLDUBSJEOG-UHFFFAOYSA-N | |
| 2734814 | |
| COC1=C(C=C(C=C1)C(=O)OC)N |
Safety and Handling
EINECSNumber : (3)-1656
TSCA : No