missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-iodobenzoate, 97%

Catalog No. AAB2569903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2569903 1 g
AAB2569906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2569903 Supplier Thermo Scientific Chemicals Supplier No. B2569903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 412947-54-7 | C8H8INO2 | 277.061 g/mol

Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.

Chemical Identifiers

CAS 412947-54-7
Molecular Formula C8H8INO2
Molecular Weight (g/mol) 277.061
MDL Number MFCD08064225
InChI Key WJEBNIVVLJEIKE-UHFFFAOYSA-N
Synonym 3-amino-4-iodobenzoic acid methyl ester, benzoic acid, 3-amino-4-iodo-, methyl ester, methyl3-amino-4-iodobenzoate, pubchem10509, acmc-209ygt, ksc495i4l, methyl-3-amino-4-iodobenzoate, methyl 3-amino-4-iodo-benzoate, 3-amino-4-iodo-benzoic acid methyl ester
PubChem CID 21628210
IUPAC Name methyl 3-amino-4-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)N

Specifications

Melting Point 134°C to 136°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 277.06
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-iodobenzoate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only