missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-2-methylbenzoate 98.0+%, TCI America™

Catalog No. A15635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15635G 5 g
A156325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15635G Supplier TCI America Supplier No. A15635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18583-89-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00211328
InChI Key ZOOQFAUFPWXUMI-UHFFFAOYSA-N
Synonym 3-amino-2-methylbenzoic acid methyl ester, methyl3-amino-2-methylbenzoate, methyl-2-methyl-3-aminobenzoate, methyl 3-amino-2-methyl-benzoate, methyl2-methyl-3-aminobenzoate, 3-amino-o-toluic acid methyl ester, benzoic acid, 3-amino-2-methyl-, methyl ester, pubchem4978, acmc-1c8sv, ksc495o4h
PubChem CID 3801025
IUPAC Name methyl 3-amino-2-methylbenzoate
SMILES CC1=C(C=CC=C1N)C(=O)OC

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl 3-Amino-2-methylbenzoate