missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dimethoxybenzoate, 98%

Catalog No. p-7081483
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1148609 10 g
AAA1148618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1148609 Supplier Thermo Scientific Chemicals Supplier No. A1148609
Only null left

CAS: 2150-37-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2150-37-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008432
InChI Key YXUIOVUOFQKWDM-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzoic acid methyl ester, benzoic acid, 3,5-dimethoxy-, methyl ester, 3,5-dimethoxybenzoic acid methyl, methyl 3,5-bis methyloxy benzoate, acmc-1ccvs, rarechem al bf 0065, methyl 3,5-dimethoxybnezoate, ksc489q8p, ethyl 3,5-dimethoxybenzoate, methyl 3,5-dimethoxybenzoate
PubChem CID 75074
IUPAC Name methyl 3,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC(OC)=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 298°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2693126
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dimethoxybenzoate
EINECSNumber 218-423-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only