missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dihydroxybenzoate 98.0+%, TCI America™

Catalog No. p-7127173
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D221525G 25 g
D2215250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D221525G Supplier TCI America Supplier No. D221525G
Only null left

Chemical Identifiers

CAS 2150-44-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002289
InChI Key RNVFYQUEEMZKLR-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzoic acid methyl ester, benzoic acid, 3,5-dihydroxy-, methyl ester, methyl3,5-dihydroxybenzoate, methyl .alpha.-resorcylate, alpha-resorcinol carboxylic acid methyl ester, alpha-resorcylic acid, methyl ester, .alpha.-resorcylic acid, methyl ester, .alpha.-resorcinol carboxylic acid methyl ester, pubchem21357, methyl alpha-resorcylate
PubChem CID 75076
IUPAC Name methyl 3,5-dihydroxybenzoate
SMILES COC(=O)C1=CC(O)=CC(O)=C1

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dihydroxybenzoate