Learn More
Methyl 3,5-dihydroxybenzoate, 98%
CAS: 2150-44-9 | C8H8O4 | 168.15 g/mol
Supplier: Thermo Scientific Chemicals A1522418
Description
Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Methyl 3,5-dihydroxybenzoate | |
| 164°C to 168°C | |
| MFCD00002289 | |
| 2091651 | |
| Slightly soluble in water. | |
| COC(=O)C1=CC(O)=CC(O)=C1 | |
| 168.15 | |
| 168.15 |
| 2150-44-9 | |
| C8H8O4 | |
| 50 g | |
| 3,5-dihydroxybenzoic acid methyl ester, benzoic acid, 3,5-dihydroxy-, methyl ester, methyl3,5-dihydroxybenzoate, methyl .alpha.-resorcylate, alpha-resorcinol carboxylic acid methyl ester, alpha-resorcylic acid, methyl ester, .alpha.-resorcylic acid, methyl ester, .alpha.-resorcinol carboxylic acid methyl ester, pubchem21357, methyl alpha-resorcylate | |
| RNVFYQUEEMZKLR-UHFFFAOYSA-N | |
| methyl 3,5-dihydroxybenzoate | |
| 75076 | |
| 98% |
Chemical Identifiers
| 2150-44-9 | |
| 168.15 | |
| RNVFYQUEEMZKLR-UHFFFAOYSA-N | |
| 75076 | |
| COC(=O)C1=CC(O)=CC(O)=C1 |
| C8H8O4 | |
| MFCD00002289 | |
| 3,5-dihydroxybenzoic acid methyl ester, benzoic acid, 3,5-dihydroxy-, methyl ester, methyl3,5-dihydroxybenzoate, methyl .alpha.-resorcylate, alpha-resorcinol carboxylic acid methyl ester, alpha-resorcylic acid, methyl ester, .alpha.-resorcylic acid, methyl ester, .alpha.-resorcinol carboxylic acid methyl ester, pubchem21357, methyl alpha-resorcylate | |
| methyl 3,5-dihydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-426-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only