missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-difluorobenzoate, 98%

Catalog No. AAL1717106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1717106 5 g
AAL1717114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1717106 Supplier Thermo Scientific Chemicals Supplier No. L1717106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 216393-55-4 | C8H6F2O2 | 172.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216393-55-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD01318160
InChI Key JBEJPGWPXIIQBB-UHFFFAOYSA-N
PubChem CID 2775360
IUPAC Name methyl 3,5-difluorobenzoate
SMILES COC(=O)C1=CC(F)=CC(F)=C1

Specifications

Refractive Index 1.474
Quantity 5 g
Synonym 3,5-difluorobenzoic acid methyl ester, benzoic acid, 3,5-difluoro-, methyl ester, acmc-209fmq, methyl-3,5-difluorophenylacetate, methyl-3,5-difluorobenzoate, 3,5-difluorobenzoicacid methyl ester, benzoic acid,3,5-difluoro-, methyl ester
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material Methyl 3,5-difluorobenzoate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only