missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-Hydroxyphenyl)propionate 98.0+%, TCI America™

Catalog No. p-7133084
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21965G 5 g
M219625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21965G Supplier TCI America Supplier No. M21965G

Chemical Identifiers

CAS 5597-50-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00071577
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
PubChem CID 79706
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 40°C
Boiling Point 108°C
Quantity 5 g
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-(4-Hydroxyphenyl)propionate
TSCA No
Recommended Storage Refrigerator