accessibility menu, dialog, popup

UserName

H3COOOH

CAS RN 5597-50-2

IUPAC Name: methyl 3-(4-hydroxyphenyl)propanoate
Synonyms: methyl 3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: XRAMJHXWXCMGJM-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=C(O)C=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)