missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-hydroxyphenyl)propionate, 98%, Thermo Scientific™

Catalog No. AC369430050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369430050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC369430050 Supplier Thermo Scientific Chemicals Supplier No. 369430050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 5597-50-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00071577
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
PubChem CID 79706
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)O

Specifications

Boiling Point 108.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2CO2CH3
Quantity 5g
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Methyl 3-(4-hydroxyphenyl)propionate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only