missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-hydroxyphenyl)propionate, 98%

Catalog No. p-7055626
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6425514 25 g
AAH6425522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6425514 Supplier Thermo Scientific Chemicals Supplier No. H6425514

CAS: 5597-50-2 | C10H12O3 | 180.203 g/mol

Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 5597-50-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00071577
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
PubChem CID 79706
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 39°C to 41°C (literature)
Color White
Boiling Point 108°C (11 mmHg)
Flash Point >113°C (235°F)
Quantity 25 g
Beilstein 2211073
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
Solubility Information Soluble in chloroform and methanol.
Formula Weight 180.2
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Methyl 3-(4-hydroxyphenyl)propionate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only