missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-Dimethoxybenzoate 98.0+%, TCI America™

Catalog No. M28055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28055G 5 g
M280525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28055G Supplier TCI America Supplier No. M28055G

Chemical Identifiers

CAS 2150-38-1
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008430
InChI Key BIGQPYZPEWAPBG-UHFFFAOYSA-N
Synonym methyl veratrate, benzoic acid, 3,4-dimethoxy-, methyl ester, 3,4-dimethoxybenzoic acid methyl ester, veratric acid, methyl ester, methylveratrate, pubchem13009, acmc-1cfay, veratric acid methyl ester, methyl 3,4 dimethoxybenzoate, methyl 3, 4-dimethoxybenzoate
PubChem CID 16522
ChEBI CHEBI:86906
IUPAC Name methyl 3,4-dimethoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 283°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4-Dimethoxybenzoate