missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-Dihydroxyphenylacetate 98.0+%, TCI America™

Catalog No. D27345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27345G 5 g
D27341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27345G Supplier TCI America Supplier No. D27345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25379-88-8
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00583539
InChI Key UGFILLIGHGZLHE-UHFFFAOYSA-N
Synonym Catechol-4-acetic Acid Methyl Ester, 3,4-Dihydroxyphenylacetic Acid Methyl Ester, Homoprotocatechuic Acid Methyl Ester, Pyrocatechol-4-acetic Acid Methyl Ester
PubChem CID 11008556
IUPAC Name methyl 2-(3,4-dihydroxyphenyl)acetate
SMILES COC(=O)CC1=CC(O)=C(O)C=C1

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4-Dihydroxyphenylacetate