missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dihydroxybenzoate, 97%

Catalog No. AAL0955204
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0955204 2 g
AAL0955209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0955204 Supplier Thermo Scientific Chemicals Supplier No. L0955204
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2150-43-8 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2150-43-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016437
InChI Key CUFLZUDASVUNOE-UHFFFAOYSA-N
Synonym methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
PubChem CID 287064
IUPAC Name methyl 3,4-dihydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 134°C to 138°C
Odor Odorless
Quantity 2 g
Beilstein 1637939
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dihydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only