missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals

$135.04 - $135.04

Chemical Identifiers

CAS 2905-68-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00018165
InChI Key WICGATIORFSOJL-UHFFFAOYSA-N
PubChem CID 76193
IUPAC Name methyl 3,4-dichlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(Cl)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2002706
View Documents
Thermo Scientific Chemicals
B2002706
5 g
Each for $135.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2905-68-2
205.03
WICGATIORFSOJL-UHFFFAOYSA-N
methyl 3,4-dichlorobenzoate
C8H6Cl2O2
MFCD00018165
76193
COC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

2905-68-2
C8H6Cl2O2
5 g
WICGATIORFSOJL-UHFFFAOYSA-N
methyl 3,4-dichlorobenzoate
76193
97%
44°C to 46°C
MFCD00018165
methyl 3,4-dichlorobenzenecarboxylate, 3,4-dichlorobenzoic acid methyl ester, benzoic acid, 3,4-dichloro-, methyl ester, benzoic acid,3,4-dichloro-, methyl ester, benzoic acid,4-dichloro-, methyl ester, acmc-1ch33, methyldichlorobenzenecarboxylate, methyl 3,4-bis chloranyl benzoate
COC(=O)C1=CC=C(Cl)C(Cl)=C1
205.03
205.04
Methyl 3,4-dichlorobenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only