missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog No. AAB2002706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2002706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2002706 Supplier Thermo Scientific Chemicals Supplier No. B2002706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-68-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00018165
InChI Key WICGATIORFSOJL-UHFFFAOYSA-N
PubChem CID 76193
IUPAC Name methyl 3,4-dichlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 44°C to 46°C
Quantity 5 g
Synonym methyl 3,4-dichlorobenzenecarboxylate, 3,4-dichlorobenzoic acid methyl ester, benzoic acid, 3,4-dichloro-, methyl ester, benzoic acid,3,4-dichloro-, methyl ester, benzoic acid,4-dichloro-, methyl ester, acmc-1ch33, methyldichlorobenzenecarboxylate, methyl 3,4-bis chloranyl benzoate
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dichlorobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only