missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M16491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16491G 1 g
M16495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16491G Supplier TCI America Supplier No. M16491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79528-48-6
Molecular Formula C21H23NO9S
Molecular Weight (g/mol) 465.473
InChI Key QNHCRERIIKDXAI-PLQGUWCTSA-N
PubChem CID 13991662
IUPAC Name [(2R,3S,4R,5R,6S)-3,4-diacetyloxy-5-(1,3-dioxoisoindol-2-yl)-6-methylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC)N2C(=O)C3=CC=CC=C3C2=O)OC(=O)C)OC(=O)C

Specifications

Melting Point 150°C
Color White
Quantity 1 g
Formula Weight 465.47
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer