accessibility menu, dialog, popup

UserName

CAS RN 79528-48-6

IUPAC Name: [3,4-bis(acetyloxy)-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-6-(methylsulfanyl)oxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-5-(1,3-dioxoisoindol-2-yl)-6-(methylsulfanyl)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 465.47
Molecular Formula: C21H23NO9S
InChi Key: QNHCRERIIKDXAI-UHFFFAOYNA-N
SMILES: CSC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results