missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-Tris(benzyloxy)benzoate 98.0+%, TCI America™

Catalog No. T18705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18705G Supplier TCI America Supplier No. T18705G

Chemical Identifiers

CAS 70424-94-1
Molecular Formula C29H26O5
Molecular Weight (g/mol) 454.522
MDL Number MFCD03425302
InChI Key MAUSJTNDKNFSEU-UHFFFAOYSA-N
Synonym Methyl 3,4,5-Tribenzyloxybenzoate, 3,4,5-Tribenzyloxybenzoic Acid Methyl Ester, 3,4,5-Tris(benzyloxy)benzoic Acid Methyl Ester
PubChem CID 352181
IUPAC Name methyl 3,4,5-tris(phenylmethoxy)benzoate
SMILES COC(=O)C1=CC(=C(C(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 102°C
Color White to Yellow
Quantity 5 g
Formula Weight 454.52
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4,5-Tris(benzyloxy)benzoate