accessibility menu, dialog, popup

UserName

CH3OOOOO

CAS RN 70424-94-1

IUPAC Name: methyl 3,4,5-tris(benzyloxy)benzoate
Synonyms: methyl 3,4,5-tris(benzyloxy)benzoate
Molecular Weight (g/mol): 454.52
Molecular Formula: C29H26O5
InChi Key: MAUSJTNDKNFSEU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OCC2=CC=CC=C2)=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results